Product Details

SNP ID
rs200598823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100707161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCACCGTGGAGCTTGTTGATGAG[C/G]TGGAGCCAGAGACCGACACACGGGA
Phenotype
MIM: 606113
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
POP7 PubMed Links

Gene Details

Gene
POP7
Gene Name
POP7 homolog, ribonuclease P/MRP subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005837.2 593 Missense Mutation CTG,GTG L111V NP_005828.2

View Full Product Details