Product Details

SNP ID
rs200072505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73570769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAAAGCACTCCTCCTTCTCGCCC[C/T]GCCGGGTATTGTAGAGATGAATACT
Phenotype
MIM: 605842
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBL2 PubMed Links

Gene Details

Gene
TBL2
Gene Name
transducin (beta)-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012453.2 1086 Missense Mutation CAG,CGG Q361R NP_036585.1
XM_006715923.3 1086 Missense Mutation CAG,CGG Q328R XP_006715986.1
XM_011516027.2 1086 Missense Mutation CAG,CGG Q196R XP_011514329.1

View Full Product Details