Product Details

SNP ID
rs201719232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:117278166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGCCTGCTGGGGTCATGTAGCGG[C/T]TGTCCAGTCAGCGTTCTTGGGGGCC
Phenotype
MIM: 147870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WNT2 PubMed Links

Gene Details

Gene
WNT2
Gene Name
Wnt family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003391.2 1372 Missense Mutation ACC,GCC T358A NP_003382.1

View Full Product Details