Product Details

SNP ID
rs201890499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:101196083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGTGGGGTGGAGGCGCTGGGGGA[C/T]GGGGATGGGGCGGCCCACTGCAGGG
Phenotype
MIM: 610184 MIM: 603066
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MOGAT3 PubMed Links

Gene Details

Gene
MOGAT3
Gene Name
monoacylglycerol O-acyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287147.1 1058 Missense Mutation NP_001274076.1
NM_178176.3 1058 Missense Mutation NP_835470.1
XM_005250309.3 1058 Silent Mutation XP_005250366.1
XM_011516126.2 1058 Missense Mutation XP_011514428.1
XM_011516127.2 1058 Missense Mutation XP_011514429.1
Gene
PLOD3
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 3
There are no transcripts associated with this gene.

View Full Product Details