Product Details

SNP ID
rs201761598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27199394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGTAGCCCTGGTGGTAGAAGGC[A/G]AACTCCTTAGCGCGGGAGCTGAACT
Phenotype
MIM: 614060 MIM: 142959
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOTTIP PubMed Links

Gene Details

Gene
HOTTIP
Gene Name
HOXA distal transcript antisense RNA
There are no transcripts associated with this gene.

Gene
HOXA13
Gene Name
homeobox A13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000522.4 713 Silent Mutation TTC,TTT F228F NP_000513.2

View Full Product Details