Product Details

SNP ID
rs201351489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27748530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTTGTTTCAGATTCACAATGAC[A/G]TCCTTTCAAGAAGTCCCATTGCAGA
Phenotype
MIM: 605326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAX1BP1 PubMed Links

Gene Details

Gene
TAX1BP1
Gene Name
Tax1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079864.2 188 Silent Mutation ACA,ACG T2T NP_001073333.1
NM_001206901.1 188 Silent Mutation ACA,ACG T2T NP_001193830.1
NM_001206902.1 188 UTR 5 NP_001193831.1
NM_006024.6 188 Silent Mutation ACA,ACG T2T NP_006015.4
XM_005249900.2 188 Silent Mutation ACA,ACG T2T XP_005249957.1
XM_017012754.1 188 Silent Mutation ACA,ACG T2T XP_016868243.1

View Full Product Details