Product Details

SNP ID
rs199775926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:77196606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGTGCCATTATAGTTACCAAC[A/G]TGTAAACGATATTTGAGAAACTCAT
Phenotype
MIM: 605351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC146 PubMed Links

Gene Details

Gene
CCDC146
Gene Name
coiled-coil domain containing 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020879.2 1033 Intron NP_065930.2
Gene
FGL2
Gene Name
fibrinogen like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006682.2 1033 Silent Mutation CAC,CAT H331H NP_006673.1

View Full Product Details