Product Details
- SNP ID
-
rs201142786
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:11974005 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCATATTTTTACAGCAAGACAGAA[C/T]ATTGTGGCAGAACGCAATCCACCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB135
PubMed Links
Gene Details
- Gene
- DEFB135
- Gene Name
- defensin beta 135
There are no transcripts associated with this gene.
- Gene
- DEFB136
- Gene Name
- defensin beta 136
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001033018.2 |
169 |
Missense Mutation |
ATT,GTT |
I57V |
NP_001028190.2 |
View Full Product Details