Product Details

SNP ID
rs201186533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38057171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCAAGGGATCTGCCCACCATTC[C/T]GGGGGTCACCAGCCCTTCCAGTGAT
Phenotype
MIM: 602223
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF4EBP1 PubMed Links

Gene Details

Gene
EIF4EBP1
Gene Name
eukaryotic translation initiation factor 4E binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004095.3 Intron NP_004086.1

View Full Product Details