Product Details

SNP ID
rs201312888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:492589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTGGCCACCCACGGTGTCCTCA[C/G]GGTCAGCCGATATGTCTTCAAGAGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TDRP PubMed Links

Gene Details

Gene
TDRP
Gene Name
testis development related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256113.1 938 Missense Mutation CCT,CGT P123R NP_001243042.1
NM_175075.4 938 Missense Mutation CCT,CGT P123R NP_778250.2
XM_006716232.3 938 Missense Mutation CCT,CGT P123R XP_006716295.1
XM_017013113.1 938 Missense Mutation CCT,CGT P133R XP_016868602.1

View Full Product Details