Product Details

SNP ID
rs201124259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:492591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGCCACCCACGGTGTCCTCAGG[G/T]TCAGCCGATATGTCTTCAAGAGCAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TDRP PubMed Links

Gene Details

Gene
TDRP
Gene Name
testis development related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256113.1 936 Missense Mutation GAA,GAC E122D NP_001243042.1
NM_175075.4 936 Missense Mutation GAA,GAC E122D NP_778250.2
XM_006716232.3 936 Missense Mutation GAA,GAC E122D XP_006716295.1
XM_017013113.1 936 Missense Mutation GAA,GAC E132D XP_016868602.1

View Full Product Details