Product Details

SNP ID
rs201010270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:54136628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGGAGGCCAGACTCCATTTTAC[A/G]TCCGAATCCCAAAATACGGGTTTAA
Phenotype
MIM: 611828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL15 PubMed Links

Gene Details

Gene
MRPL15
Gene Name
mitochondrial ribosomal protein L15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014175.3 300 Missense Mutation ATC,GTC I76V NP_054894.1
XM_017013362.1 300 Missense Mutation ATC,GTC I76V XP_016868851.1

View Full Product Details