Product Details
- SNP ID
-
rs201094510
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:6924824 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTCTCTGTTCTCATCCCTCAGAG[A/G]CAGCAGAATCTGTGGTTAATACCCA
- Phenotype
-
MIM: 600471
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFA6
PubMed Links
Gene Details
- Gene
- DEFA6
- Gene Name
- defensin alpha 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001926.3 |
338 |
Silent Mutation |
TGC,TGT |
C99C |
NP_001917.1 |
View Full Product Details