Product Details

SNP ID
rs201858328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11984498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCAAGGTACTTGCCGGCCAAAA[C/T]GTCTAAAAAACGAACAATATCGTAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DEFB134 PubMed Links

Gene Details

Gene
DEFB134
Gene Name
defensin beta 134
There are no transcripts associated with this gene.

Gene
DEFB135
Gene Name
defensin beta 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033017.2 142 Missense Mutation CGT,TGT R48C NP_001028189.2
Gene
DEFB136
Gene Name
defensin beta 136
There are no transcripts associated with this gene.

View Full Product Details