Product Details

SNP ID
rs199977411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:93922737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTATAGACCTCAACACTGGTGA[C/G]TCGACTGATATTGATGTTAAGGAGG
Phenotype
MIM: 605993
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR378D2 PubMed Links

Gene Details

Gene
MIR378D2
Gene Name
microRNA 378d-2
There are no transcripts associated with this gene.

Gene
PDP1
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 1
There are no transcripts associated with this gene.

View Full Product Details