Product Details

SNP ID
rs199916173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142613685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCCGTCCTCGCCGGGGACCCAC[C/G]GCTGGTACTGCTGGGCCTCGGCGGG
Phenotype
MIM: 612461
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARC PubMed Links

Gene Details

Gene
ARC
Gene Name
activity regulated cytoskeleton associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015193.4 788 Missense Mutation CCG,CGG P196R NP_056008.1

View Full Product Details