Product Details

SNP ID
rs200259637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:26507754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAAAGGAGGCCTCTTCCTCCTC[C/T]TCTTCCCGTATTACCTTCATTAGCT
Phenotype
MIM: 602463 MIM: 603970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPYSL2 PubMed Links

Gene Details

Gene
DPYSL2
Gene Name
dihydropyrimidinase like 2
There are no transcripts associated with this gene.

Gene
PNMA2
Gene Name
paraneoplastic Ma antigen 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007257.5 1772 Silent Mutation GAA,GAG E334E NP_009188.1
XM_011544365.2 1772 Silent Mutation GAA,GAG E334E XP_011542667.1

View Full Product Details