Product Details

SNP ID
rs199866548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:116848952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACAACAGCGGCAATACCTGAAG[A/G]CCATGAAGCATCTGCTGAGTCCTTT
Phenotype
MIM: 606462
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD21 PubMed Links

Gene Details

Gene
RAD21
Gene Name
RAD21 cohesin complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006265.2 1986 Silent Mutation GGC,GGT G566G NP_006256.1

View Full Product Details