Product Details

SNP ID
rs201265114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137582596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGACGCTTCCTCTCCCGACAGT[A/G]CTTCTTGTGGGCAGGCCAGTCCTTC
Phenotype
MIM: 613210 MIM: 611424
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPH7 PubMed Links

Gene Details

Gene
DPH7
Gene Name
diphthamide biosynthesis 7
There are no transcripts associated with this gene.

Gene
ZMYND19
Gene Name
zinc finger MYND-type containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138462.2 853 Missense Mutation CAC,TAC H211Y NP_612471.1
XM_005266052.4 853 Missense Mutation CAC,TAC H272Y XP_005266109.1
XM_017014255.1 853 Intron XP_016869744.1

View Full Product Details