Product Details
- SNP ID
-
rs200387267
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:114337183 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCCCGGCTCAGGGCCTTGTTGAG[C/T]TCCTCTAGGTCGCCCAGGTCGAGCT
- Phenotype
-
MIM: 605729
MIM: 138610
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AKNA
PubMed Links
Gene Details
- Gene
- AKNA
- Gene Name
- AT-hook transcription factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001317950.1 |
3996 |
Silent Mutation |
GAA,GAG |
E1397E |
NP_001304879.1 |
NM_001317952.1 |
3996 |
Silent Mutation |
GAA,GAG |
E1278E |
NP_001304881.1 |
NM_030767.5 |
3996 |
Silent Mutation |
GAA,GAG |
E1397E |
NP_110394.3 |
XM_005252244.2 |
3996 |
Silent Mutation |
GAA,GAG |
E1397E |
XP_005252301.1 |
XM_005252245.1 |
3996 |
Silent Mutation |
GAA,GAG |
E1397E |
XP_005252302.1 |
XM_005252247.4 |
3996 |
Silent Mutation |
GAA,GAG |
E1397E |
XP_005252304.1 |
XM_005252248.2 |
3996 |
Silent Mutation |
GAA,GAG |
E1365E |
XP_005252305.1 |
XM_006717294.1 |
3996 |
Silent Mutation |
GAA,GAG |
E1397E |
XP_006717357.1 |
XM_006717295.2 |
3996 |
Intron |
|
|
XP_006717358.1 |
XM_011519063.2 |
3996 |
Silent Mutation |
GAA,GAG |
E1278E |
XP_011517365.1 |
XM_011519064.2 |
3996 |
Intron |
|
|
XP_011517366.1 |
XM_011519065.2 |
3996 |
Silent Mutation |
GAA,GAG |
E1278E |
XP_011517367.2 |
XM_011519066.2 |
3996 |
Intron |
|
|
XP_011517368.1 |
XM_017015172.1 |
3996 |
Silent Mutation |
GAA,GAG |
E1278E |
XP_016870661.1 |
- Gene
- ORM2
- Gene Name
- orosomucoid 2
There are no transcripts associated with this gene.
View Full Product Details