Product Details

SNP ID
rs200456325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35075566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCCAGAGTGGGCAGTATGGCAG[T/G]TCCTTGGTTCCTTTTCTGGCATCTT
Phenotype
MIM: 602956 MIM: 614730 MIM: 601023
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
FANCG PubMed Links

Gene Details

Gene
FANCG
Gene Name
Fanconi anemia complementation group G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004629.1 1824 Missense Mutation GAA,GAC E444D NP_004620.1
Gene
PIGO
Gene Name
phosphatidylinositol glycan anchor biosynthesis class O
There are no transcripts associated with this gene.

Gene
VCP
Gene Name
valosin containing protein
There are no transcripts associated with this gene.

View Full Product Details