Product Details
- SNP ID
-
rs201383603
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:135624271 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATAATAGAAATCTCCCTGTCCAAA[C/T]GGGATGCAAGCTGCTGAGGTCGGCC
- Phenotype
-
MIM: 613699
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GLT6D1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17040344] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GLT6D1
- Gene Name
- glycosyltransferase 6 domain containing 1
- Gene
- LOC102723971
- Gene Name
- lipocalin 1-like
There are no transcripts associated with this gene.
View Full Product Details