Product Details

SNP ID
rs200709450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:111686771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAACATTGCTCTTCTCCTCCTT[A/C]AAAAAAACCTCAGCCGAGTCTGCCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C9orf84 PubMed Links

Gene Details

Gene
C9orf84
Gene Name
chromosome 9 open reading frame 84
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011518309.1 Intron XP_011516611.1
XM_017014341.1 Intron XP_016869830.1

View Full Product Details