Product Details
- SNP ID
-
rs200460186
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:2221212 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACTGCTGCGTAGATGGAAGTCCA[C/T]GCTCCTTCATCGGGGGTCTGGTGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PubMed Links
Gene Details
View Full Product Details