Product Details

SNP ID
rs199728999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113276670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCAAAGGAGAGTCTGGGATTTT[C/G]GGGAAAGACGGACTTAAAGCAGGGA
Phenotype
MIM: 614533 MIM: 607795
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDC26 PubMed Links

Gene Details

Gene
CDC26
Gene Name
cell division cycle 26
There are no transcripts associated with this gene.

Gene
PRPF4
Gene Name
pre-mRNA processing factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244926.1 263 Missense Mutation TTC,TTG F50L NP_001231855.1
NM_001322266.1 263 UTR 5 NP_001309195.1
NM_001322267.1 263 UTR 5 NP_001309196.1
NM_004697.4 263 Missense Mutation TTC,TTG F51L NP_004688.2

View Full Product Details