Product Details

SNP ID
rs200111116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113275751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCTCCAGAGCCCAGCATGGCTT[C/T]CTCGCGAGCCTCTTCCACGGTACAG
Phenotype
MIM: 614533 MIM: 607795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC26 PubMed Links

Gene Details

Gene
CDC26
Gene Name
cell division cycle 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139286.3 118 Intron NP_644815.1
XM_017014573.1 118 Intron XP_016870062.1
XM_017014574.1 118 Intron XP_016870063.1
XM_017014575.1 118 Intron XP_016870064.1
Gene
PRPF4
Gene Name
pre-mRNA processing factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244926.1 118 Missense Mutation TCC,TTC S3F NP_001231855.1
NM_001322266.1 118 UTR 5 NP_001309195.1
NM_001322267.1 118 UTR 5 NP_001309196.1
NM_004697.4 118 Missense Mutation TCC,TTC S3F NP_004688.2

View Full Product Details