Product Details
- SNP ID
-
rs201598978
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:27524356 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAAATGAGCACCAAACCTGATA[G/T]GATTCAAAAGTGTTTGTGGCTTGAG
- Phenotype
-
MIM: 615326
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IFNK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144060127,rs56087834] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IFNK
- Gene Name
- interferon kappa
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020124.2 |
43 |
Missense Mutation |
AGG,ATG |
R7M |
NP_064509.2 |
- Gene
- MOB3B
- Gene Name
- MOB kinase activator 3B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024761.4 |
43 |
Intron |
|
|
NP_079037.3 |
View Full Product Details