Product Details
- SNP ID
-
rs201919906
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:90613255 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATGATCACGCACTTGCCTTTGAG[C/T]TTCTCTTTCCTTTTCTGCTGCTTGC
- Phenotype
-
MIM: 607863
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DIRAS2
PubMed Links
Gene Details
- Gene
- DIRAS2
- Gene Name
- DIRAS family GTPase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017594.4 |
963 |
Silent Mutation |
AAA,AAG |
K191K |
NP_060064.2 |
View Full Product Details