Product Details
- SNP ID
-
rs199822985
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:76428899 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATCGAGCACCTCCGTGCCGCCCAC[C/T]CCTGGTGGGGGACTGAGCACCTCCG
- Phenotype
-
MIM: 300759
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAGEE1
PubMed Links
Gene Details
- Gene
- MAGEE1
- Gene Name
- MAGE family member E1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020932.2 |
1247 |
Silent Mutation |
ACC,ACT |
T323T |
NP_065983.1 |
View Full Product Details