Product Details

SNP ID
rs202162113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49830445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGCAGCGATGAGTGCACGAGTGA[G/T]ATCAAGATCCAGAGGAAGAGGAGAT
Phenotype
MIM: 300287
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PAGE4 PubMed Links

Gene Details

Gene
PAGE4
Gene Name
PAGE family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318877.1 96 Missense Mutation AGA,ATA R6I NP_001305806.1
NM_007003.4 96 Missense Mutation AGA,ATA R6I NP_008934.1

View Full Product Details