Product Details

SNP ID
rs202163127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49270627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGCTGCCCTTTGCTGAGGGCG[C/T]GGGCGATGGGGCGCGCCTCGACTTC
Phenotype
MIM: 300292
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXP3 PubMed Links

Gene Details

Gene
FOXP3
Gene Name
forkhead box P3
There are no transcripts associated with this gene.

Gene
PPP1R3F
Gene Name
protein phosphatase 1 regulatory subunit 3F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184745.1 785 Intron NP_001171674.1
NM_033215.4 785 Missense Mutation GCG,GTG A253V NP_149992.3
XM_005272687.3 785 Missense Mutation GCG,GTG A253V XP_005272744.1
XM_017029932.1 785 Intron XP_016885421.1
XM_017029933.1 785 Intron XP_016885422.1
XM_017029934.1 785 Missense Mutation GCG,GTG A253V XP_016885423.1
XM_017029935.1 785 Missense Mutation GCG,GTG A253V XP_016885424.1

View Full Product Details