Product Details

SNP ID
rs200904268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:21857433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCCCTTTCAGTGCACATTCGAA[G/T]GCTGCGGGAAACGCTTTTCCCTTGA
Phenotype
MIM: 300294 MIM: 300570
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MBTPS2 PubMed Links

Gene Details

Gene
MBTPS2
Gene Name
membrane bound transcription factor peptidase, site 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015884.3 1447 Intron NP_056968.1
Gene
YY2
Gene Name
YY2 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206923.3 1447 Missense Mutation GGC,TGC G317C NP_996806.2

View Full Product Details