Product Details

SNP ID
rs201301539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:78272925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACAAGGTTATGACCACGGACTT[C/T]TGCATTCTAAGGACAGAATCACAGG
Phenotype
MIM: 300201
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYSLTR1 PubMed Links

Gene Details

Gene
CYSLTR1
Gene Name
cysteinyl leukotriene receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282186.1 1236 Silent Mutation CAA,CAG Q274Q NP_001269115.1
NM_001282187.1 1236 Silent Mutation CAA,CAG Q274Q NP_001269116.1
NM_001282188.1 1236 Silent Mutation CAA,CAG Q274Q NP_001269117.1
NM_006639.3 1236 Silent Mutation CAA,CAG Q274Q NP_006630.1

View Full Product Details