Product Details

SNP ID
rs201575340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:121048245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGGGTGCTAAAGCTGGTGTTAA[A/G]ATCAATCCCAAGAACTATACCGAAA
Phenotype
MIM: 300144
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLUD2 PubMed Links

Gene Details

Gene
GLUD2
Gene Name
glutamate dehydrogenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012084.3 Intron NP_036216.2

View Full Product Details