Product Details

SNP ID
rs201497020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49174350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCCCCAGGTACTGGGCCTAATC[A/G]CTACGTGCCTCTTTGGCTATGATGC
Phenotype
MIM: 300112 MIM: 300111
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAGIX PubMed Links

Gene Details

Gene
MAGIX
Gene Name
MAGI family member, X-linked
There are no transcripts associated with this gene.

Gene
PLP2
Gene Name
proteolipid protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002668.2 525 Missense Mutation ACT,GCT T121A NP_002659.1
Gene
PRICKLE3
Gene Name
prickle planar cell polarity protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307979.1 525 Intron NP_001294908.1
NM_006150.4 525 Intron NP_006141.2

View Full Product Details