Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001199148.1 | 2100 | Intron | NP_001186077.1 | ||
NM_173512.2 | 2100 | Intron | NP_775783.1 | ||
XM_005246350.4 | 2100 | Intron | XP_005246407.1 | ||
XM_006712337.3 | 2100 | Intron | XP_006712400.1 | ||
XM_011510737.2 | 2100 | Intron | XP_011509039.1 | ||
XM_011510742.2 | 2100 | Intron | XP_011509044.1 | ||
XM_017003455.1 | 2100 | Intron | XP_016858944.1 | ||
XM_017003456.1 | 2100 | Intron | XP_016858945.1 | ||
XM_017003457.1 | 2100 | Intron | XP_016858946.1 | ||
XM_017003458.1 | 2100 | Intron | XP_016858947.1 | ||
XM_017003459.1 | 2100 | Intron | XP_016858948.1 | ||
XM_017003460.1 | 2100 | Intron | XP_016858949.1 | ||
XM_017003461.1 | 2100 | Intron | XP_016858950.1 | ||
XM_017003462.1 | 2100 | UTR 3 | XP_016858951.1 | ||
XM_017003463.1 | 2100 | Intron | XP_016858952.1 | ||
XM_017003464.1 | 2100 | Intron | XP_016858953.1 | ||
XM_017003465.1 | 2100 | Intron | XP_016858954.1 |