Product Details
- SNP ID
-
rs6118469
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:9075173 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCATCCCAGGATGTTAAAGTTAGAA[C/T]TGAGACTGAAACTCAGGTAGGCTGA
- Phenotype
-
MIM: 600810
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PLCB4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74391622] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PLCB4
- Gene Name
- phospholipase C beta 4
View Full Product Details