Product Details

SNP ID
rs11831881
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106360852 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTTATAGGCTTATAGAGAGAGAC[A/G]TGTAAGCAAGTGGAATAACATGCAA
Phenotype
MIM: 614366
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
POLR3B PubMed Links

Gene Details

Gene
POLR3B
Gene Name
RNA polymerase III subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160708.1 Intron NP_001154180.1
NM_018082.5 Intron NP_060552.4
XM_017019621.1 Intron XP_016875110.1

View Full Product Details