Product Details

SNP ID
rs2058157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:22899461 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTAATTAAGTTGTAAGTCATCAT[A/G]AGACTGACCAACCTGTCCCTCTGCA
Phenotype
MIM: 610531
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM126A PubMed Links

Gene Details

Gene
FAM126A
Gene Name
family with sequence similarity 126 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032581.3 Intron NP_115970.2
XM_005249894.3 Intron XP_005249951.1
XM_005249895.2 Intron XP_005249952.1
XM_011515589.2 Intron XP_011513891.1
XM_011515590.1 Intron XP_011513892.1

View Full Product Details