Product Details

SNP ID
rs3828191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:164493392 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTGATCCCTGAGAAGCACTGCTA[C/T]TCTACTTCATCTAGTTCTAATTCTC
Phenotype
MIM: 601524
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRB14 PubMed Links

Gene Details

Gene
GRB14
Gene Name
growth factor receptor bound protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303422.1 Intron NP_001290351.1
NM_004490.2 Intron NP_004481.2
XM_011511022.2 Intron XP_011509324.1
XM_017003899.1 Intron XP_016859388.1

View Full Product Details