Product Details

SNP ID
rs6122877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50127259 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCATAGCAGTTAGCACCATTAG[C/T]CACGCTACACATTTTATTCATTCGT
Phenotype
MIM: 610994
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM189 PubMed Links

Gene Details

Gene
TMEM189
Gene Name
transmembrane protein 189
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162505.1 Intron NP_001155977.1
NM_199129.2 Intron NP_954580.1
Gene
TMEM189-UBE2V1
Gene Name
TMEM189-UBE2V1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199203.2 Intron NP_954673.1

View Full Product Details