Product Details

SNP ID
rs9405048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30702515 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCCAGCCACCTCTTTAACTGCCA[C/T]TGCCACCTCACCCAGACCCAGAACA
Phenotype
MIM: 607593
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MDC1 PubMed Links

Gene Details

Gene
MDC1
Gene Name
mediator of DNA damage checkpoint 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014641.2 Intron NP_055456.2
XM_005249492.2 Intron XP_005249549.1
XM_005249493.3 Intron XP_005249550.1
XM_005249494.4 Intron XP_005249551.1
XM_005249497.3 Intron XP_005249554.1
XM_005249498.3 Intron XP_005249555.1
XM_011515001.2 Intron XP_011513303.1
XM_011515003.2 Intron XP_011513305.1
XM_011515004.2 Intron XP_011513306.1
XM_017011519.1 Intron XP_016867008.1
XM_017011520.1 Intron XP_016867009.1
XM_017011521.1 Intron XP_016867010.1
XM_017011522.1 Intron XP_016867011.1
Gene
MDC1-AS1
Gene Name
MDC1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details