Product Details

SNP ID
rs12572757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:113753136 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGTAAACACTGCTTTAAAATAT[A/G]CCACCCACAGCTAATCTCTCCTGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLEKHS1 PubMed Links

Gene Details

Gene
PLEKHS1
Gene Name
pleckstrin homology domain containing S1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193434.1 Intron NP_001180363.1
NM_001193435.1 Intron NP_001180364.1
NM_024889.4 Intron NP_079165.3
NM_182601.1 Intron NP_872407.1
XM_005270162.2 Intron XP_005270219.1
XM_005270163.2 Intron XP_005270220.1
XM_011540171.1 Intron XP_011538473.1
XM_011540172.1 Intron XP_011538474.1
XM_011540173.1 Intron XP_011538475.1
XM_011540174.2 Intron XP_011538476.1
XM_011540175.1 Intron XP_011538477.1
XM_011540176.1 Intron XP_011538478.1
XM_017016666.1 Intron XP_016872155.1

View Full Product Details