Product Details
- SNP ID
-
rs2909028
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:148094 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACACAAATGAGAGCATTCTGTAT[C/T]GTTTCATTTCTATGAAGGTGAAAAG
- Phenotype
-
MIM: 142310
MIM: 600928
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HBZ
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117806895] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HBZ
- Gene Name
- hemoglobin subunit zeta
- Gene
- NPRL3
- Gene Name
- NPR3 like, GATOR1 complex subunit
There are no transcripts associated with this gene.
View Full Product Details