Product Details

SNP ID
rs35728395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:143171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGGCAGGATAGGAGGCCTTATC[A/G]GTCCTGCCACTTGGTCTGGGCCTCA
Phenotype
MIM: 142310 MIM: 600928
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HBZ PubMed Links

Gene Details

Gene
HBZ
Gene Name
hemoglobin subunit zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005332.2 Intron NP_005323.1
XM_005255287.3 Intron XP_005255344.1
XM_005255288.3 Intron XP_005255345.1
Gene
NPRL3
Gene Name
NPR3 like, GATOR1 complex subunit
There are no transcripts associated with this gene.

View Full Product Details