Product Details

SNP ID
rs6138991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3043424 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTAAAGCAAGAGTCCTGGGCCTG[A/G]AGGCTCCTGGGAAGGTGGTGGGGAG
Phenotype
MIM: 602352 MIM: 611988 MIM: 176884
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNRH2 PubMed Links

Gene Details

Gene
GNRH2
Gene Name
gonadotropin releasing hormone 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310220.1 1668 Intron NP_001297149.1
NM_001501.1 1668 Intron NP_001492.1
NM_178331.1 1668 Intron NP_847901.1
NM_178332.1 1668 Intron NP_847902.1
XM_017027823.1 1668 UTR 5 XP_016883312.1
Gene
MRPS26
Gene Name
mitochondrial ribosomal protein S26
There are no transcripts associated with this gene.

Gene
PTPRA
Gene Name
protein tyrosine phosphatase, receptor type A
There are no transcripts associated with this gene.

View Full Product Details