Product Details

SNP ID
rs7743050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167325088 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGACCCTGCATCATCAGCAACCAG[C/T]GCTCCCTCCAGCCTCCGCGCATTTC
Phenotype
MIM: 607995
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTLL2 PubMed Links

Gene Details

Gene
TTLL2
Gene Name
tubulin tyrosine ligase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031949.4 3 UTR 5 NP_114155.4
XM_006715572.3 3 Intron XP_006715635.1
Gene
UNC93A
Gene Name
unc-93 homolog A (C. elegans)
There are no transcripts associated with this gene.

View Full Product Details