Product Details

SNP ID
rs3800962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:18026946 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATTTTGGAGCAATGCTTCATCTT[A/C]TCATCTTGAGGTATGGTATTGGTGA
Phenotype
MIM: 611566
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRPS1L1 PubMed Links

Gene Details

Gene
PRPS1L1
Gene Name
phosphoribosyl pyrophosphate synthetase 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175886.2 918 Missense Mutation GAG,GAT E279D NP_787082.1

View Full Product Details