Product Details

SNP ID
rs12667934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:20617335 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTATCATTCAGTACATGTGGGGCA[A/C]GGCCTAGGTAGGTGTCTGATATCTT
Phenotype
MIM: 611785
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ABCB5 PubMed Links

Gene Details

Gene
ABCB5
Gene Name
ATP binding cassette subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163941.1 Intron NP_001157413.1
NM_001163942.1 Intron NP_001157414.1
NM_001163993.2 Intron NP_001157465.1
NM_178559.5 Intron NP_848654.3
XM_011515367.2 Intron XP_011513669.1

View Full Product Details