Product Details

SNP ID
rs13280716
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:96270332 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CATGTGGACTCTGCAGCCAAGTTCT[A/G]TTTCATGATTAACTATGGAAACTTC
Phenotype
MIM: 616930 MIM: 612792
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTERF3 PubMed Links

Gene Details

Gene
MTERF3
Gene Name
mitochondrial transcription termination factor 3
There are no transcripts associated with this gene.

Gene
PTDSS1
Gene Name
phosphatidylserine synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290225.1 Intron NP_001277154.1
NM_014754.2 Intron NP_055569.1

View Full Product Details